3-(4-Chloro-2-isopropylphenoxy)azetidine

Supplier: Matrix Scientific

049421-500MG 049421-2.5G
101887-588EA 364.62 USD
101887-588 101887-590
3-(4-Chloro-2-isopropylphenoxy)azetidine
3-(4-Chloro-2-isopropylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₂H₁₆ClNO
MDL Number: MFCD13559890
CAS Number: 1220027-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR