4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole ≥97%
Supplier: Matrix Scientific
002397-1G
002397-5G
101783-834EA
24.62
USD
101783-834
101783-836
4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole ≥97%
4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole
Hazard: Irritant
Formula:
C₅H₄ClF₃N₂ |
MDL Number:
MFCD00155724 CAS Number: 235106-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...