2-Ethylpyrrole ≥98%

Supplier: Matrix Scientific

006166-1G 006166-5G
101790-184EA 67.69 USD
101790-184 101790-186
2-Ethylpyrrole ≥98%
2-Ethylpyrrole

Hazard: Irritant/Stored under Argon

Formula: C₆H₉N
Boiling Pt: 69…72 °C (20 mmHg)
Density: 0.92
MDL Number: MFCD00060477
CAS Number: 1551-06-0

Order Now

Specification Test Results

Boiling point 69-72°C/20m
Density 0.92


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR