Hexadecafluorosebaconitrile ≥97%

Supplier: Matrix Scientific

Synonyms: Perfluorosebaconitrile

006191-1G 006191-5G 006191-25G
101790-222EA 75.38 USD
101790-222 101790-224 101790-226
Hexadecafluorosebaconitrile ≥97%
Hexadecafluorosebaconitrile

Hazard: Irritant-Harmful

Formula: C₁₀F₁₆N₂
MW: 452.09 g/mol
Boiling Pt: 147…148 °C
Density: 1.673
MDL Number: MFCD00042366
CAS Number: 2342-09-8

Order Now

Specification Test Results

Boiling point 147-148°C
Density 1.673
ND20 1.3039


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR