Ethyl-2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate

Supplier: Matrix Scientific

067162-500MG 067162-1G
101933-348EA 201.54 USD
101933-348 101933-350
Ethyl-2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate
Ethyl-2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₉H₈ClN₃O₂S
MW: 257.7 g/mol
Melting Pt: 253…255 °C
MDL Number: MFCD18074287
CAS Number: 847560-46-7

Order Now

Specification Test Results

Melting point 253-255°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR