1-[2-(Thiomorpholin-4-yl)ethyl]imidazolidin-2-one ≥95%
Supplier: Matrix Scientific
066897-500MG
066897-1G
101932-682EA
600
USD
101932-682
101932-684
1-[2-(Thiomorpholin-4-yl)ethyl]imidazolidin-2-one ≥95%
1-[2-(Thiomorpholin-4-yl)ethyl]imidazolidin-2-one
Hazard: Irritant
Formula:
C₉H₁₇N₃OS Melting Pt: 129…131 °C |
MDL Number:
MFCD19982779 CAS Number: 1331823-96-1 |
Specification Test Results
Melting point | 129-131°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...