Methyl-3-[(hydroxyimino)methyl]-1H-indole-2-carboxylate ≥95%
Supplier: Matrix Scientific
043979-500MG
043979-1G
043979-5G
101863-494EA
278.46
USD
101863-494
101863-496
101863-498
Methyl-3-[(hydroxyimino)methyl]-1H-indole-2-carboxylate ≥95%
Methyl-3-[(hydroxyimino)methyl]-1H-indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₀N₂O₃ MW: 218.21 g/mol Melting Pt: 206…208 °C |
MDL Number:
MFCD04125006 CAS Number: 861211-41-8 |
Specification Test Results
Melting point | 206-208°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...