6-Amino-2H-1,4-benzoxazin-3(4H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Amino-3,4-dihydro-3-oxo-2H-1,4-benzoxazine,6-Amino-3-oxo-3,4-dihydrobenzo[1,4]oxazine,6-Amino-2H-benzo[b][1,4]oxazin-3(4H)-one,6-Amino-4H-benzo[1,4]oxazin-3-one, 6-amino-2,4-dihydro-1,4-benzoxazin-3-one
043926-1G
043926-5G
101863-244EA
76.92
USD
101863-244
101863-246
6-Amino-2H-1,4-benzoxazin-3(4H)-one ≥95%
6-Amino-2H-1,4-benzoxazin-3(4H)-one
Hazard: Irritant
Formula:
C₈H₈N₂O₂ MW: 164.17 g/mol Melting Pt: 266…268 °C |
MDL Number:
MFCD03425742 CAS Number: 89976-75-0 |
Specification Test Results
Melting point | 266-268°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...