2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(9,10-Dioxo-9,10-dihydroanthracen-2-yl)acetonitrile

043638-500MG 043638-1G 043638-5G
101861-820EA 747.69 USD
101861-820 101861-822 101861-824
2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetonitrile ≥95%
2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetonitrile

Hazard: Irritant

Formula: C₁₆H₉NO₂
MW: 247.26 g/mol
Melting Pt: 215…217 °C
MDL Number: MFCD01315214
CAS Number: 121831-04-7

Order Now

Specification Test Results

Melting point 215-217°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR