4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether ≥95%

Supplier: Matrix Scientific

043232-500MG 043232-1G 043232-5G
101859-812EA 375.38 USD
101859-812 101859-814 101859-816
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether ≥95%
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether

Hazard: Irritant

Formula: C₁₄H₉F₃N₂S
Melting Pt: 78…80 °C
MDL Number: MFCD00793554
CAS Number: 320421-51-0

Order Now

Specification Test Results

Melting point 78-80°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR