6-Chlorouracil

Supplier: Matrix Scientific

Synonyms: 6-Chloro-2,4-dihydroxypyrimidine, 6-Chloropyrimidine-2,4(1H,3H)-dione, 4-Chlorouracil, 4-(3-Bromophenyl)pyrazole, 4-Chloro-2,6-dihydroxypyrimidine, 6-Chloro-2,4(1H,3H)-pyrimidinedione

006834-5G 006834-25G
101791-266EA 15.38 USD
101791-266 101791-268
6-Chlorouracil
6-Chlorouracil

Hazard: Irritant

Formula: C₄H₃ClN₂O₂
Melting Pt: 301…302 °C (dec.)
MDL Number: MFCD00014595
CAS Number: 4270-27-3

Order Now

Specification Test Results

Melting point 301-302°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR