5-Fluoroorotic acid monohydrate (5-FOA) ≥98%

Supplier: Matrix Scientific

Synonyms: 5-FOA, 5-Fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-Fluorouracil-6-carboxylic acid monohydrate

006158-5G 006158-25G
101790-164EA 284.62 USD
101790-164 101790-166
5-Fluoroorotic acid monohydrate (5-FOA) ≥98%
5-Fluoroorotic acid monohydrate

Hazard: Irritant/Store in Freezer

Formula: C₅H₃FN₂O₄·1H₂O
MW: 174.0 g/mol (anhy)
Melting Pt: 278 °C
MDL Number: MFCD00150658
CAS Number: 220141-70-8

Order Now

Specification Test Results

Melting point 278°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR