2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile ≥96%
Supplier: Matrix Scientific
Synonyms:
2-(2,2,2-Trifluoroethoxy)nicotinonitrile
002878-250MG
002878-1G
101784-828EA
21.54
USD
101784-828
101784-830
2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile ≥96%
2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile
Purity: 96%
CAS: 175277-89-1
MDL Number: MFCD00275529
Molecular Formula: C8H5F3N2O
Molecular Weight: 202,14
Hazard: Irritant
Formula:
C₈H₅F₃N₂O Melting Pt: 44…46 °C |
MDL Number:
MFCD00275529 CAS Number: 175277-89-1 |
Specification Test Results
Melting point | 44-46°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...