3-[(4-Methylbenzyl)oxy]-2-thiophenecarbohydrazide ≥95%

Supplier: Matrix Scientific

045233-500MG 045233-1G 045233-5G
101869-828EA 375.38 USD
101869-828 101869-830 101869-832
3-[(4-Methylbenzyl)oxy]-2-thiophenecarbohydrazide ≥95%
3-[(4-Methylbenzyl)oxy]-2-thiophenecarbohydrazide

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂S
Melting Pt: 134…136 °C
MDL Number: MFCD00215167
CAS Number: 343375-82-6

Order Now

Specification Test Results

Melting point 134-136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR