1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Amino-2,2,2-trifluoroacetophenone, 1-(4-Aminophenyl)-2,2,2-trifluoroethanone

044507-1G 044507-5G 044507-250MG
101866-238EA 280 USD
101866-238 101866-240 102663-716
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone ≥95%
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone

Hazard: Irritant

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD05663790
CAS Number: 23516-79-2

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR