Cyclopropyl methyl ketone ≥98% for synthesis
Supplier: Matrix Scientific
|
Danger
|
Synonyms:
1-Cyclopropylethanone
067246-100G
067246-250G
101933-628EA
75.38
USD
101933-628
101933-630
Cyclopropyl methyl ketone ≥98% for synthesis
Cyclopropyl methyl ketone
Hazard code(s): Flammable, Toxic
Formula:
C₅H₈O Boiling Pt: 111…113 °C Melting Pt: >–70 °C Density: 0.898 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00001297 CAS Number: 765-43-5 EINECS: 212-146-4 UN: 1992 ADR: 3,II |
Specification Test Results
Boiling point | 111-113°C |
Melting point |
|
Density | 0.898 |
ND20 | 1.424 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...