4-Fluorophenyl isocyanate for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene

038319-500MG
101845-264EA 146.15 USD
101845-264
4-Fluorophenyl isocyanate for synthesis
4-Fluorophenyl isocyanate
Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 42…47 °C (1 mmHg)
Density: 1.206 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002023
CAS Number: 1195-45-5
EINECS: 214-799-0
UN: 3080
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR