Indole-7-carboxylic acid ≥98%

Supplier: Matrix Scientific

024707-5G 024707-25G
101818-698EA 112.31 USD
101818-698 101818-700
Indole-7-carboxylic acid ≥98%
Indole-7-carboxylic acid

Hazard code(s): Irritant

Formula: C₉H₇NO₂
MDL Number: MFCD00210442
CAS Number: 1670-83-3
EINECS: 216-801-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR