6-Methoxy-m-toluidine

Supplier: Matrix Scientific
Danger

Synonyms: 2-Methoxy-5-methylaniline1-Amino-2-methoxy-5-methylbenzenep-Cresidine5-Methyl-o-anisidine2-Amino-4-methylanisole3-Amino-4-methoxytoluene

022113-500MG
101813-388EA 315.38 USD
101813-388
6-Methoxy-m-toluidine
6-Methoxy-m-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 55.5 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00007815
CAS Number: 120-71-8
EINECS: 204-419-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR