4-Bromo-m-cresol ≥95%
Supplier: Matrix Scientific
|
Warning
|
016032-5G
016032-25G
016032-100G
101803-940EA
15.38
USD
101803-940
101803-942
101803-944
4-Bromo-m-cresol ≥95%
4-Bromo-m-cresol
Hazard code(s): Irritant
Formula:
C₇H₇BrO Boiling Pt: 142…145 °C (23mm) Melting Pt: 59…61 °C |
MDL Number:
MFCD00079723 CAS Number: 14472-14-1 EINECS: 238-464-3 |
Specification Test Results
Boiling point | 142-145°C/2 |
Melting point | 59-61°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...