3-Bromo-4-fluoroaniline ≥98% for synthesis

Supplier: Matrix Scientific

012817-25G 012817-100G
101803-448EA 44.62 USD
101803-448 101803-450
3-Bromo-4-fluoroaniline ≥98% for synthesis
3-Bromo-4-fluoroaniline

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03788559
CAS Number: 656-64-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR