4-Hydroxythiophenol ≥97%

Supplier: Matrix Scientific

012524-5G 012524-25G
101802-754EA 52.31 USD
101802-754 101802-756
4-Hydroxythiophenol ≥97%
4-Hydroxythiophenol

Hazard code(s): Irritant-Harmful, Stench, Air Sensitive, Refrigerate

Formula: C₆H₆OS
Boiling Pt: 149…150 °C (25mm)
Melting Pt: 33…35 °C
MDL Number: MFCD00004850
CAS Number: 637-89-8
EINECS: 211-307-6

Order Now

Specification Test Results

Boiling point 149-150°C/2
Melting point 33-35°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR