3-Cyano-4-methylpyridine ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile
011111-5G
011111-25G
101799-884EA
36.92
USD
101799-884
101799-886
3-Cyano-4-methylpyridine ≥98% for synthesis
3-Cyano-4-methylpyridine
Hazard code(s): Irritant, Refrigerate
Formula:
C₇H₆N₂ Boiling Pt: 108…111 °C (20mm) Melting Pt: 41…44 °C |
MDL Number:
MFCD00234272 CAS Number: 5444-01-9 |
Specification Test Results
Boiling point | 108-111°C/2 |
Melting point | 41-44°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...