4-Bromobiphenyl (PBB No. 3) ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl
011104-25G
011104-100G
101799-860EA
15.38
USD
101799-860
101799-862
4-Bromobiphenyl (PBB No. 3) ≥98%
4-Bromobiphenyl (PBB No. 3)
Hazard code(s): Irritant
Formula:
C₁₂H₉Br MW: 233.11 g/mol Boiling Pt: 310 °C Melting Pt: 90…92 °C Density: 1.615 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000100 CAS Number: 92-66-0 EINECS: 202-176-6 |
Specification Test Results
Boiling point | 310°C |
Melting point | 90-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...