1,4-Dibromonaphthalene ≥98% for synthesis

Supplier: Matrix Scientific
Warning

007550-5G 007550-25G
101793-024EA 38.46 USD
101793-024 101793-026
1,4-Dibromonaphthalene ≥98% for synthesis
1,4-Dibromonaphthalene

Hazard code(s): Irritant

Formula: C₁₀H₆Br₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 80…82 °C
MDL Number: MFCD00041823
CAS Number: 83-53-4
EINECS: 201-484-8

Order Now

Specification Test Results

Melting point 80-82°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR