3-Methylanthranil acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
007451-25G
007451-100G
101792-798EA
18.46
USD
101792-798
101792-800
3-Methylanthranil acid ≥97%
3-Methylanthranil acid
Hazard code(s): Irritant
Formula:
C₈H₉NO₂ Boiling Pt: 315 °C (1013 hPa) Melting Pt: 176…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007745 CAS Number: 4389-45-1 EINECS: 224-505-2 |
Specification Test Results
Melting point | 176-178°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...