4'-Hydroxychalcone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(4-Hydroxyphenyl)-3-phenylprop-2-en-1-one, 4'-Hydroxybenzylidenacetophenone

006583-5G 006583-25G
101790-792EA 21.54 USD
101790-792 101790-794
4'-Hydroxychalcone ≥97%
4'-Hydroxychalcone

Hazard code(s): Irritant

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 175…177 °C
MDL Number: MFCD00016484
CAS Number: 2657-25-2

Order Now

Specification Test Results

Melting point 175-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR