Tris(4-fluorophenyl)phosphine ≥98%

Supplier: Matrix Scientific

006501-1G
101790-654EA 44.62 USD
101790-654
Tris(4-fluorophenyl)phosphine ≥98%
Tris(4-fluorophenyl)phosphine

Hazard code(s): Irritant

Formula: C₁₈H₁₂F₃P
MW: 316.26 g/mol
Melting Pt: 80…81 °C
MDL Number: MFCD00013553
CAS Number: 18437-78-0

Order Now

Specification Test Results

Melting point 80-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR