Chloropentafluorobenzene ≥99% for synthesis

Supplier: Matrix Scientific

Synonyms: Pentafluorochlorobenzene

006261-25G 006261-100G 006261-500G
101790-286EA 36.92 USD
101790-286 101790-288 101790-290
Chloropentafluorobenzene ≥99% for synthesis
Chloropentafluorobenzene

Hazard code(s): Irritant

Formula: C₆ClF₅
Boiling Pt: 122…123 °C (750mm)
Melting Pt: –15 °C
Density: 1.568 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000534
CAS Number: 344-07-0
EINECS: 206-450-6

Order Now

Specification Test Results

Boiling point 122-123°C/7
Density 1.568
ND20 1.423


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR