1,2-Diiodotetrafluorobenzene ≥99%

Supplier: Matrix Scientific
Warning

005636-1G 005636-5G
101789-268EA 61.54 USD
101789-268 101789-270
1,2-Diiodotetrafluorobenzene ≥99%
1,2-Diiodotetrafluorobenzene

Hazard code(s): Irritant, Refrigerate

Formula: C₆F₄I₂
MW: 401.87 g/mol
Melting Pt: 49…50 °C
MDL Number: MFCD00019011
CAS Number: 2708-97-6
EINECS: 220-303-3

Order Now

Specification Test Results

Melting point 49-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR