3,4-Dibromothiophene ≥98% for synthesis

Supplier: Matrix Scientific
Warning

004598-25G 004598-100G
101787-562EA 21.54 USD
101787-562 101787-564
3,4-Dibromothiophene ≥98% for synthesis
3,4-Dibromothiophene

Hazard code(s): Irritant

Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221
Melting Pt: 4.5
Density: 2.188 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8

Order Now

Specification Test Results

Boiling point 221-222°C
Melting point 4-5°C
Density 2.188
ND20 1.64


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR