(4-Fluorophenyl)acetic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-(4-Fluorophenyl)acetic acid

003890-100G 003890-250G
101786-372EA 53.85 USD
101786-372 101786-374
(4-Fluorophenyl)acetic acid ≥98% for synthesis
(4-Fluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄F)CH₂COOH
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 83…85 °C
MDL Number: MFCD00004343
CAS Number: 405-50-5
EINECS: 206-972-4
Merck Index: 13,04204

Order Now

Specification Test Results

Melting point 83-85°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR