3'-Fluoroacetophenone ≥99%
Supplier: Matrix Scientific
|
Warning
|
002922-25G
002922-100G
101784-918EA
69.23
USD
101784-918
101784-920
3'-Fluoroacetophenone ≥99%
3'-Fluoroacetophenone
Hazard code(s): Irritant
Formula:
C₈H₇FO Boiling Pt: 81 °C (9mm) Melting Pt: –3 °C Density: 1.126 g/cm³ |
MDL Number:
MFCD00000338 CAS Number: 455-36-7 EINECS: 207-245-4 |
Specification Test Results
Boiling point | 81°C/9mm |
Density | 1.126 |
ND20 | 1.509 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...