4-Fluoro-3-hydroxybenzoic acid ≥97% for synthesis

Supplier: Matrix Scientific

002354-5G 002354-25G 002354-100G
101783-726EA 76.92 USD
101783-726 101783-728 101783-730
4-Fluoro-3-hydroxybenzoic acid ≥97% for synthesis
4-Fluoro-3-hydroxybenzoic acid

Hazard code(s): Irritant

Formula: C₇H₅FO₃
MDL Number: MFCD00269641
CAS Number: 51446-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR