(3-Bromo-4-methoxyphenyl)acetic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-methoxyphenylacetic acid, 2-(3-Bromo-4-methoxyphenyl)acetic acid

001796-5G 001796-25G
101782-600EA 35.38 USD
101782-600 101782-602
(3-Bromo-4-methoxyphenyl)acetic acid ≥98% for synthesis
(3-Bromo-4-methoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₉H₉BrO₃
Melting Pt: 115…117 °C
MDL Number: MFCD00017538
CAS Number: 774-81-2

Order Now

Specification Test Results

Melting point 115-117°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR