Nitric acid 1.00 N

Supplier: Ricca Chemical
Danger

Synonyms: Nitric acid solution

R5430000-1A R5430000-20F R5430000-2.5D R5430000-4A R5430000-500A
RC543032EA 86 USD
RC543032 RC5430-5 RC5430-5PT RCR5430000-4A RCR5430000500A
Nitric acid 1.00 N
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Density: 1.035 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Tris(hydroxymethyl)aminomethane/Methyl Red) 0.999-1.001 N at 20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR