Sulfuric acid 12.0 N in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R8342000-4A
R8342000-1A
R8342000-500A
R8342000-20F
RC8342-1EA
351
USD
RC8342-1
RC8342-32
RC8342-16
RC8342-5
Sulfuric acid 12.0 N in aqueous solution
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 12.00 ± 0.05 N at 20.0 °C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...