Oxalic acid 0.0500 mol/L (0.100 N) volumetric solution

Supplier: Ricca Chemical

Synonyms: Ethanedioic acid

R5471100-500A R5471100-1A
RC5471.1-16EA 61 USD
RC5471.1-16 RC5471.1-32
Oxalic acid 0.0500 mol/L (0.100 N) volumetric solution
Oxalic acid
Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
Merck Index: 13,06980

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 40) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR