An Ideal Organism for Studying Vertebrate Embryology and Development.
- Observe the whole zebrafish life cycle in approximately a week.
- Transparent embryos allow observation during development
After the fish hatch, students can continue to raise the fish until adulthood and start the process all over again. Not only will students learn about embryology, they will also learn about photoperiodism since mating is induced by controlling a specific light/dark cycle, as well as how to culture Paramecium and brine shrimp to provide a constant food supply for the developing fry.
Ordering information: Materials in the Basic Kit include Netted fish breeding chamber, Dip net, Brine shrimp egg hatchery, Light control timer, Instructions, and Coupon redeemable for 4 female and 2 male Zebrafish and a culture of Paramecium caudatum. Materials in the Complete Kit include all materials in Basic Kit, plus 5 1/2 gallon glass aquarium with hinged glass top and light, Air pump and filter, Heater, and thermometer.
Materials not included but required are low power magnification (available separately) and additional aquarium (available separately) to keep the adults from cannibalizing their young.
This item contains a coupon for live or perishable materials. At checkout, please provide a date that the live or perishable components will arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight freight, additional charges will apply.