Ward's® Live Shigella flexneri Culture - PATHOGEN
Supplier: Avantor
470179-152EA 13.95 USD
470179-152 470179-558
Ward's® Live Shigella flexneri Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
From sewage and human intestine. Causes dysentery. 
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Negative
  • Morphology: Rods
  • Gram Reaction: Negative
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
These organisms are offered as ready-to-use tube cultures, as well as in freeze-dried cultures for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This organism is pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.