Ward's® Chemistry Endospore Stain Kit
Supplier: ALDON CORP SE
SS0922-PK/2 SS0922-PK/2A
470233-558EA 14.39 USD
470233-558 470233-560
Ward's® Chemistry Endospore Stain Kit
Educational Classroom Kits and Activities
Distinguish between endospores and the vegetative structures of bacterial cells. The endospores will stain green against a pink background. Includes spore staining solution, safranin O solution, and instructions.
Shelf life is 3 years.