Safety Shower and Eye/Face Wash, Speakman®
Supplier: SPEAKMAN COMPANY SE
HFO SE-603 SE-623
470121-230EA 171.95 USD
470121-230 470204-068 470204-070
Safety Shower and Eye/Face Wash, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Combination shower with rectangular eye/face wash bowl.
  • Stainless steel bowl
  • 8" diameter yellow plastic showerhead
  • 3 year limited warranty
  • Hand and foot operation optional
This combinated safety device can be used to flush contaminants from eyes, face, or entire body. Space saving, stainless steel unit consists of a shower and eye wash with stainless steel basin. The shower has a 20.3 cm (8") diameter head with impeller action and stay-open valve. The eye/face wash is actuated by a stainless steel push handle. Six aerated outlets in each spray head provide a 22.8×20.3 cm (9×8") curtain of aerated water for more extensive washing action. Monthly flushing is recommended.
Certifications: Meet ANSI/ISEA Z358.1.