Potassium Sulfate

Supplier: ALDON CORP SE

PP0785-500ML PP0770-100G PP0770-500GP PP0770-100GR PP0770-500GR PP0781-500ML
470302-268EA 13.5 USD
470302-268 470302-262 470302-266 470302-270 470302-272 470225-712
Potassium Sulfate
Chemicals for Science Education

CAS Number: 7778-80-5
Formula Weight: 174.27
Formula: K2SO4
Density (g/mL): 2.662
Boiling Point (°C): 1067
Freezing Point (°C): 1689
Solubility: Water
Synonyms: Sal Polychrestum
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR