Potassium Carbonate Anhydrous

Supplier: ALDON CORP SE

PP0420-500G PP0420-500GR PP0431-500ML PP0420-2.5KG
470302-068EA 27.2 USD
470302-068 470302-070 470225-664 470230-896
Potassium Carbonate Anhydrous

CAS Number: 584-08-7
Formula: K2CO3
Density: 2.428 g/mL
Freezing Point: 891°C
Solubility: Water
Synonyms: Potash, Carbonic Acid Dipotassium Salt
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR