Polyvinyl Alcohol

Supplier: ALDON CORP SE

PP0117-1L PP0118-500G PP0117-250ML PP0119-100ML PP0119-3.8L PP0119-1L PP0118-100G
470302-030EA 24.25 USD
470302-030 470302-040 470302-028 470302-032 470302-034 470302-036 470302-038
Polyvinyl Alcohol
Chemicals for Science Education

CAS Number: 9002-89-5
Formula: CH2CHOHn
Synonyms: PVA, Ethanol Homopolymer
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR