Ward's® Plant Mineral Deficiency Lab Activity
Supplier: Ward's Science
CC02663KIT
470024-820EA 47.99 USD
470024-820 470021-146
Ward's® Plant Mineral Deficiency Lab Activity
Educational Classroom Kits and Activities
Combine Hydroponics and the Study of Mineral Deficiencies
With this WARD’S exclusive activity, your students germinate two types of seeds, then transfer them to various nutrient solutions for observation. Within two weeks, your students will become experts at identifying the symptoms of calcium, phosphorus, sulfur, magnesium, iron, nitrogen, and potassium soil deficiencies, all grown hydroponically. You will get a teacher’s manual and enough materials for ten setups.