Lysozyme

Supplier: ALDON CORP SE

LL0280-1GR
470301-618EA 63.9 USD
470301-618
Lysozyme
Chemicals for Science Education

CAS Number: 9001-63-2
Formula: Complex Enzyme
Boiling Point (°C): Decomposes
Freezing Point (°C): stable <55
Synonyms: Globulin G, Muramidase
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR