LaMotte® Portable Water Analysis Laboratory
Supplier: LaMotte
1991-02 R-1991-02
470004-280EA 3692 USD
470004-280 470004-282
LaMotte® Portable Water Analysis Laboratory
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Comprehensive Outfit Includes Digital Colorimeter
Using an electronic digital colorimeter is more reliable and quicker than standard eye comparisons to color charts.
Tests and battery-operated equipment for 20 factors are all included in a rugged case. Colorimetric tests are ammonia, bromine, chlorine, chromium, copper, fluoride, iodine, iron, nitrate, nitrite, phosphate (low), silica, sulfate, sulfide, and turbidity.
Direct reading titrator tests are included for alkalinity, carbon dioxide, chloride/salinity, dissolved oxygen, and hardness. Sufficient materials for 50 tests per factor are included.
Note: Contains sodium arsenite and barium compounds