Ward's® Kocuria rosea
Supplier: Avantor
470176-372EA 13.95 USD
470176-372 470176-374
Ward's® Kocuria rosea
Specimens Live Specimens Bacteria - Living Material
This organism is commonly isolated from soil or water, and produces a pink or rose colored pigment.
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Cocci in tetrads
  • Gram Stain: Positive
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.