Freeze-Dried Bullfrogs
Supplier: Avantor
470027-850PK 204.95 USD
470027-850 470001-404
Freeze-Dried Bullfrogs
Specimens Preserved Specimens Preserved Animal Materials
Freeze-dried bullfrogs. Available in vacuum packs.
  • Amphibian anatomy
  • Economical
Our freeze-dried specimens, which use no chemicals, can be rehydrated in 10% alcohol for dissection. Size: 5" and over.